Learn More
Medchemexpress LLC Sodium zirconium cyclosilicate | 17141-74-1 | 95.0% | 25 MG
SDP
Catalog No. 5000233124
Encompass_Preferred
Sodium zirconium cyclosilicate is an inorganic, microporous zirconium silicate that acts as a selective cation exchanger for monovalent ions and is used in laboratory research on potassium binding and renal disease.
  • Selective binding of monovalent cations, especially potassium.
  • Microporous crystalline inorganic material, insoluble and non-absorbed.
  • Suitable for preclinical research on hyperkalemia and chronic kidney disease.
  • Supplied in small, accurately weighed pack sizes for laboratory use.
  • Characterized by CAS 17141-74-1 and documented in the supplier safety data sheet.
Catalog No. 50-002-33124 Supplier Medchemexpress LLC Supplier No. HY11957225MG
Sodium zirconium cyclosilicate is an inorganic, microporous zirconium silicate that acts as a selective cation exchanger for monovalent ions and is used in laboratory research on potassium binding and renal disease.
  • Selective binding of monovalent cations, especially potassium.
  • Microporous crystalline inorganic material, insoluble and non-absorbed.
  • Suitable for preclinical research on hyperkalemia and chronic kidney disease.
  • Supplied in small, accurately weighed pack sizes for laboratory use.
  • Characterized by CAS 17141-74-1 and documented in the supplier safety data sheet.