Learn More
Medchemexpress LLC Sodium zirconium cyclosilicate | 17141-74-1 | 95.0% | 5 MG
SDP
Catalog No. 5000234619
Encompass_Preferred
Sodium zirconium cyclosilicate is an inorganic, microporous zirconium silicate used as a selective cation exchanger and potassium binder for laboratory research. Supplied as a white to off-white solid in small milligram quantities, it is intended for in vitro ion-exchange and potassium regulation studies.
  • White to off-white solid form, insoluble and non-absorbed
  • Highly selective potassium binding for ion-exchange research
  • Available in small milligram quantities for laboratory use
  • Store sealed at 4°C and protect from moisture
  • Purity typically around 95.0% for research applications
Catalog No. 50-002-34619 Supplier Medchemexpress LLC Supplier No. HY1195725MG
Sodium zirconium cyclosilicate is an inorganic, microporous zirconium silicate used as a selective cation exchanger and potassium binder for laboratory research. Supplied as a white to off-white solid in small milligram quantities, it is intended for in vitro ion-exchange and potassium regulation studies.
  • White to off-white solid form, insoluble and non-absorbed
  • Highly selective potassium binding for ion-exchange research
  • Available in small milligram quantities for laboratory use
  • Store sealed at 4°C and protect from moisture
  • Purity typically around 95.0% for research applications