Learn More
Medchemexpress LLC Sodium zirconium cyclosilicate | 17141-74-1 | 95.0% | 50 MG
SDP
Catalog No. 5000232993
Encompass_Preferred
Sodium zirconium cyclosilicate is an inorganic, microporous zirconium silicate cation exchanger used in research to selectively remove excess potassium (K+) in vivo. It is supplied for preclinical and biochemical studies, including hyperkalemia and chronic kidney disease research, and is provided at laboratory-grade purity.
  • Selective potassium binding for ion-exchange studies.
  • Non-absorbed inorganic sorbent suitable for in vivo models.
  • Supplied at 95.0% purity for consistent results.
  • Available in small research pack sizes from 5 mg to 500 mg.
  • Useful for biochemical assays and renal disease research.
Catalog No. 50-002-32993 Supplier Medchemexpress LLC Supplier No. HY11957250MG
Sodium zirconium cyclosilicate is an inorganic, microporous zirconium silicate cation exchanger used in research to selectively remove excess potassium (K+) in vivo. It is supplied for preclinical and biochemical studies, including hyperkalemia and chronic kidney disease research, and is provided at laboratory-grade purity.
  • Selective potassium binding for ion-exchange studies.
  • Non-absorbed inorganic sorbent suitable for in vivo models.
  • Supplied at 95.0% purity for consistent results.
  • Available in small research pack sizes from 5 mg to 500 mg.
  • Useful for biochemical assays and renal disease research.