Learn More
Quidel Sofia 2 Campylobacter Fluorescent Immunoassay (FIA)
SDP
Catalog No. 23044181 Shop All QuidelOrtho Products
Change view
Click to view available options
Sterility:
Sterile
Catalog No. Sterility
23-044-181 Sterile
1 options
This item is not returnable. View return policy
Catalog No. 23-044-181 Supplier Quidel Supplier No. 20385
Add to Cart
Add to Cart
This item is not returnable. View return policy
An immunofluorescence-based lateral-flow technology to provide rapid qualitative detection of a Campylobacter-specific antigen in human fecal specimens.
  • Results in 15 minutes
  • Detects four of the most prevalent Campylobacter species,C. jejuni, C. coli, C. lari and C. upsaliensis
  • Can be performed with fresh, frozen, Cary Blair or C and S samples or specimens
  • Simple easy to use workflow
  • Room temperature storage
  • LIS Connectivity and Data Management: Reduced time and opportunity for human error
Specifications
Sterility Sterile
Description Sofia 2 Campylobacter tests
Storage Requirements Room temperature (15°C to 30°C)