Learn More
Quidel Sofia™ 2 RSV Test Starter Kit
SDP
Catalog No. 23043091 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
3 x 25 Tests
Catalog No. Quantity
23-043-091 3 x 25 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-043-091 Supplier Quidel Supplier No. 20313
Order eligibility is based upon executed contractual agreement. Please reach out to your Fisher HealthCare Account Representative for more information.
This item is not returnable. View return policy
Sofia 2 Instrument and RSV Kit for rapid qualitative detection of respiratory syncytial virus (RSV) nucleoprotein antigens.
Quidel™ Sofia™ 2 Fluorescent Immunoassay Analyzer
  • Small bench top analyzer that uses advanced fluorescence detection with an ultraviolet LED energy source. Sofia 2 Fluorescent Immunoassay Analyzer collects hundreds of data points on the test strip contained inside of a Sofia FIA Test Cassette.
Quidel™ Sofia™ RSV FIA Immunofluorescence Kit (x3)
  • For rapid qualitative detection of respiratory syncytial virus (RSV) nucleoprotein antigens
  • Includes: Individually packaged cassettes 25, reagent tubes 25, reagent solution 25, sterile nasopharyngeal swabs 25, large, pink fixed volume pipettes 25, small, clear fixed volume pipettes 25, RSV positive control swab 1, negative control swab 1, package insert 1, quick reference instructions 1, QC card, printer Paper 1
  • Quantity: 3 x 25 Tests
  • Test time: >15 min
  • Clia Complexity: Waived
  • Detectable Analytes : Respiratory syncytial virus nucleoprotein Antigens
  • Sample Type: Nasopharyngeal swab and nasopharyngeal aspirate/wash specimens
Order Info
Order eligibility is based upon executed contractual agreement. Please reach out to your Fisher Healthcare Account Representative for more information.
Specifications
Type Starter Kit
Includes Sofia 2 Instrument (23-043-086) and 3 Sofia RSV test kits (23-043-080).
Quantity 3 x 25 Tests