Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quidel Sofia 2 SARS Antigen+ FIA, 25 Tests
SDP

Catalog No. 23044183 Shop All QuidelOrtho Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-044-183 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-044-183 Supplier Quidel Supplier No. 20405
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The next generation of COVID-19 rapid testing, Sofia 2 SARS Antigen+ Fluorescent Immunoassay (FIA), uses advanced immunofluorescence-based lateral flow technology in a sandwich design for qualitative detection of nucleocapsid protein from SARS-CoV-2.

  • Results in 10 minutes
  • Rapid results to support efficient dispositioning of patients
  • Objective, accurate results
  • Reliable results without cross-reactivity to seasonal coronaviruses
  • Dual work modes
  • Adjusts to your volume fluctuations. Allows for significant throughput and batching of samples in READ NOW Mode
  • Virena connection
  • Provides automated tracking, data capture, government reporting, and exclusive disease mapping
  • Fluorescent technology with automated read
  • Objective result that eliminates the subjectivity of a visual read
  • All necessary components included in kit
  • Ready for use with Sofia 2 for nasal swab procedure
  • Self-contained Test Cassette
  • Clean, easy to use and dispose of

Specifications

Control Sets Positive and Negative
CE Marker Yes
Clia Complexity Waived
Detectable Analytes SARS-CoV-2
Detection Method Lateral Flow Technology
Format Cassette
Sample Type Nasal Swab
Sensitivity PPA: 89%; NPA: 99.6%
Type Rapid Test
Disposable Single-use
For Use With (Application) Fluorescence Immunoassay (FIA)
No. of Reactions 25 Tests
Quantity 25 Tests
Sterility Sterile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.