Learn More
Medchemexpress LLC HY-15005 10mg Medchemexpress, Sofosbuvir CAS:1190307-88-0 Purity:>98%
SDP
Catalog No. 502029078
Encompass_Preferred
Medchemexpress, HY-15005 10mg Sofosbuvir CAS:1190307-88-0 Sofosbuvir (GS-7977) is an HCV RNA replication inhibitor with an EC50 of 92 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9078 Supplier Medchemexpress LLC Supplier No. HY1500510MG
Medchemexpress, HY-15005 10mg Sofosbuvir CAS:1190307-88-0 Sofosbuvir (GS-7977) is an HCV RNA replication inhibitor with an EC50 of 92 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.