Learn More
Medchemexpress LLC HY-15005 1g , Sofosbuvir CAS:1190307-88-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY150051G
Encompass_Preferred
HY-15005 1g Sofosbuvir CAS: 1190307-88-0 Sofosbuvir (GS-7977) is an HCV RNA replication inhibitor with an EC50 of 92 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9527