Learn More
TARGETMOL CHEMICALS INC SOFOSBUVIR 500MG
SDP
Catalog No. 502579309
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sofosbuvir (GS 7977) is a uridine monophosphate analog inhibitor of hepatitis C virus (HCV) polymerase NS5B that is used as an antiviral agent in the treatment of chronic hepatitis C. purity: 99%
Catalog No. 50-257-9309 Supplier TARGETMOL CHEMICALS INC Supplier No. T6676500MG
Also available in 5 mg 10 mg 50 mg 100 mg 1000 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sofosbuvir (GS 7977) is a uridine monophosphate analog inhibitor of hepatitis C virus (HCV) polymerase NS5B that is used as an antiviral agent in the treatment of chronic hepatitis C. purity: 99%