Learn More
Medchemexpress LLC Sofosbuvir-d6 | 1868135-06-1 | 98.07% | 535.49 | 2 MG
SDP
Catalog No. 5000361266
Encompass_Preferred
Sofosbuvir-d6 is the deuterium-labeled Sofosbuvir, an active inhibitor of HCV RNA replication. It demonstrates potent anti-hepatitis C virus (HCV) activity and is intended for research use only.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated for drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-61266 Supplier Medchemexpress LLC Supplier No. HY15005S12MG
Sofosbuvir-d6 is the deuterium-labeled Sofosbuvir, an active inhibitor of HCV RNA replication. It demonstrates potent anti-hepatitis C virus (HCV) activity and is intended for research use only.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes incorporated for drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs