Learn More
Medchemexpress LLC HY-15005C 1mg Medchemexpress, Sofosbuvir impurity A CAS:1496552-16-9 Purity:>98%
SDP
Catalog No. 501932719
Encompass_Preferred
Medchemexpress, HY-15005C 1mg Sofosbuvir impurity A CAS:1496552-16-9 Sofosbuvir impurity A, an diastereoisomer of sofosbuvir, is the impurity of sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2719 Supplier Medchemexpress LLC Supplier No. HY15005C1MG
Medchemexpress, HY-15005C 1mg Sofosbuvir impurity A CAS:1496552-16-9 Sofosbuvir impurity A, an diastereoisomer of sofosbuvir, is the impurity of sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.