Learn More
Medchemexpress LLC HY-I0719 1mg Medchemexpress, Sofosbuvir impurity B CAS: Purity:>98%
SDP
Catalog No. 501933264
Encompass_Preferred
Medchemexpress, HY-I0719 1mg Sofosbuvir impurity B CAS: Sofosbuvir impurity B is the less active impurity of Sofosbuvir, Sofosbuvir is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrates potent anti-hepatitis C virus ( HCV ) activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3264 Supplier Medchemexpress LLC Supplier No. HYI07191MG
Medchemexpress, HY-I0719 1mg Sofosbuvir impurity B CAS: Sofosbuvir impurity B is the less active impurity of Sofosbuvir, Sofosbuvir is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrates potent anti-hepatitis C virus ( HCV ) activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.