Learn More
Medchemexpress LLC HY-I0513 1mg Medchemexpress, Sofosbuvir impurity N CAS:1394157-34-6 Purity:>98%
SDP
Catalog No. 501933258
Encompass_Preferred
Medchemexpress, HY-I0513 1mg Sofosbuvir impurity N CAS:1394157-34-6 Sofosbuvir impurity N, an diastereoisomer of sofosbuvir, is the impurity of sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3258 Supplier Medchemexpress LLC Supplier No. HYI05131MG
Medchemexpress, HY-I0513 1mg Sofosbuvir impurity N CAS:1394157-34-6 Sofosbuvir impurity N, an diastereoisomer of sofosbuvir, is the impurity of sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.