Learn More
MSC Bell Laboratories Soft Bait, 16 lb, Bromadiolone, Single-Feed
Catalog No. 502838376
Encompass
This soft bait consists of palatable bait sachets designed to eliminate rats and mice in both indoor and outdoor environments. It features bromadiolone, a second-generation anticoagulant, in a single-feed formulation. The bait comes in 10g sachets, formulated with a unique mixture of fats and oils for high palatability, and packaged to preserve the bait.
  • Palatable bait sachets
  • Single-feed formulation
  • Contains bromadiolone, a second-generation anticoagulant
  • Formulated with a unique mixture of fats and oils for high palatability
  • Sachets packaged to prevent freezing, melting, or molding
  • Ensures maximum contact between soft bait and sachet for optimal bait acceptance
  • 10g soft bait sachets
Catalog No. 50-283-8376 Supplier MSC Supplier No. 74357427
This soft bait consists of palatable bait sachets designed to eliminate rats and mice in both indoor and outdoor environments. It features bromadiolone, a second-generation anticoagulant, in a single-feed formulation. The bait comes in 10g sachets, formulated with a unique mixture of fats and oils for high palatability, and packaged to preserve the bait.
  • Palatable bait sachets
  • Single-feed formulation
  • Contains bromadiolone, a second-generation anticoagulant
  • Formulated with a unique mixture of fats and oils for high palatability
  • Sachets packaged to prevent freezing, melting, or molding
  • Ensures maximum contact between soft bait and sachet for optimal bait acceptance
  • 10g soft bait sachets