Learn More
MSC Liphatech Rodenticide Soft Bait, Bromethalin, 4 lb
Catalog No. 502838352
Encompass
Liphatech TakeDown II is an acute rodenticide soft bait formulated with Bromethalin, designed to provide rapid control of rodent populations. It is effective against House Mice, Norway Rats, and Roof Rats, including those resistant to anticoagulant rodenticides. A lethal dose can be consumed in one night's feeding, with effects appearing within 2 or more days.
  • Acute rodenticide, offering faster results than anticoagulant alternatives
  • Effective against anticoagulant-resistant house mice, Norway rats, and roof rats
  • Soft bait remains palatable and maintains integrity in hot environments
  • Packaged in convenient 4 lb bags
Catalog No. 50-283-8352 Supplier MSC Supplier No. 74359266
Add to Cart
Add to Cart
Liphatech TakeDown II is an acute rodenticide soft bait formulated with Bromethalin, designed to provide rapid control of rodent populations. It is effective against House Mice, Norway Rats, and Roof Rats, including those resistant to anticoagulant rodenticides. A lethal dose can be consumed in one night's feeding, with effects appearing within 2 or more days.
  • Acute rodenticide, offering faster results than anticoagulant alternatives
  • Effective against anticoagulant-resistant house mice, Norway rats, and roof rats
  • Soft bait remains palatable and maintains integrity in hot environments
  • Packaged in convenient 4 lb bags