Learn More
Polysciences, Inc. Soft Block, 1 gal
SDP
Catalog No. NC0181569
Encompass_Preferred
This ready-to use solution is effective on bone, nails and soft tissue. Simply soak pre-trimmed block surface in Soft Block for 5-15 minutes (depending on tissue sample size) then cut quality sections. Soft Block is environmentally safe, eliminating the need for any special waste disposal. Safe for equipment.
Catalog No. NC0181569 Supplier Polysciences, Inc. Supplier No. 246161
This ready-to use solution is effective on bone, nails and soft tissue. Simply soak pre-trimmed block surface in Soft Block for 5-15 minutes (depending on tissue sample size) then cut quality sections. Soft Block is environmentally safe, eliminating the need for any special waste disposal. Safe for equipment.