Learn More
Polysciences, Inc. Soft Block, 500 mL
SDP
Catalog No. NC9840965
Encompass
This ready-to use solution is effective on bone, nails and soft tissue. Simply soak pre-trimmed block surface in Soft Block for 5-15 minutes (depending on tissue sample size) then cut quality sections.
Catalog No. NC9840965 Supplier Polysciences, Inc. Supplier No. 24616500
This ready-to use solution is effective on bone, nails and soft tissue. Simply soak pre-trimmed block surface in Soft Block for 5-15 minutes (depending on tissue sample size) then cut quality sections.