Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Soft Drink Analysis Standard, 5 Component, Liquid, Calibration

Catalog No. 50201492 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Soft Drink Analysis Standard is a liquid calibration standard containing five components: caffeine, benzoate, sorbate, acesulfame-K, and aspartame. It is designed for use in soft drink analysis with reversed-phase separation and UV detection methods.

  • Designed for reversed phase separation.
  • Utilizes UV detection method.
  • Liquid formulation for ease of use.
  • Can be stored at room temperature.
  • Compatible with LC system types.
  • Quantitative analysis capability.
  • Shelf life of 12 months.
  • Shelf life upon reconstitution: 1 week.
  • Ships under ambient conditions.
  • Type: calibration standard.
  • Application in food analysis.
  • Contains caffeine, benzoate, sorbate, acesulfame-K, and aspartame.
  • Units per package: 1.

Catalog No. 50-201-492 Supplier Waters Corp Supplier No. 186003856
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Soft Drink Analysis Standard is a liquid calibration standard containing five components: caffeine, benzoate, sorbate, acesulfame-K, and aspartame. It is designed for use in soft drink analysis with reversed-phase separation and UV detection methods.

  • Designed for reversed phase separation.
  • Utilizes UV detection method.
  • Liquid formulation for ease of use.
  • Can be stored at room temperature.
  • Compatible with LC system types.
  • Quantitative analysis capability.
  • Shelf life of 12 months.
  • Shelf life upon reconstitution: 1 week.
  • Ships under ambient conditions.
  • Type: calibration standard.
  • Application in food analysis.
  • Contains caffeine, benzoate, sorbate, acesulfame-K, and aspartame.
  • Units per package: 1.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.