Learn More
Grainger American Hammer Soft-Face Sledge Hammer, Lead Alloy, Aluminum Handle, 10 lb Head, 1 ea
Catalog No. NC2740943
Encompass_Preferred
These lead alloy soft-face sledge hammers are specifically designed to protect critical surfaces from marring or deformation during striking applications. Ideal for tasks involving moving, adjusting, or assembling metal components, they utilize lead's unique dead blow characteristics to effectively shift material without causing surface damage. The tool features a durable aluminum handle and is suited for various maintenance and assembly tasks, such as straightening drive shafts or gear box assembly.
  • Head weight: 10 lb
  • Face diameter: 2-1/2 in.
  • Overall length: 13-1/2 in.
  • Handle material: Aluminum
  • Head material: Lead alloy
  • Non-marring design
Catalog No. NC2740943 Supplier Grainger Supplier No. 21YU65
These lead alloy soft-face sledge hammers are specifically designed to protect critical surfaces from marring or deformation during striking applications. Ideal for tasks involving moving, adjusting, or assembling metal components, they utilize lead's unique dead blow characteristics to effectively shift material without causing surface damage. The tool features a durable aluminum handle and is suited for various maintenance and assembly tasks, such as straightening drive shafts or gear box assembly.
  • Head weight: 10 lb
  • Face diameter: 2-1/2 in.
  • Overall length: 13-1/2 in.
  • Handle material: Aluminum
  • Head material: Lead alloy
  • Non-marring design