Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ Soft Scrubs™ Pants

Catalog No. 19472437 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-472-437 X-Large
19-472-453 Small
19-472-449 Medium
19-472-436 Large
19-472-433 2X-Large
19-472-434 3X-Large
19-472-435 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-472-437 Supplier International Enviroguard™ Supplier No. 2052BXL
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Soft and comfortable SMS pants feature an elastic waistband for a secure fit. Single-use scrubs offer modesty with complete coverage and can also be worn underneath gowns.

  • Scrub pants offer comfort and safety to work long hours
  • Soft pants offer modesty with complete ''no show through'' coverage
  • Can easily be worn as a protective undergarment beneath a sterile gowns
  • Quickly and easily dispose after each use to reduce spread of germs, bioburdens, and other contagions
  • Elastic waistband offers a snug, secure fit
  • Hip pocket allows storage of eyeglasses, small instruments, pens, and more
  • Possible use areas include pharmaceuticals, labs, healthcare, and controlled environments
  • Anti-static fabric is made of an opaque material that doesn't show through and resists abrasions
  • Convenient fabric loop for easily hanging up clothing
  • Reinforced crotch resists tears

Specifications

Size X-Large
Gender Unisex
Material SMS
Color Blue
Ankle Style Open
Seam Type Serged
No. of Pockets 1
Disposable Single-use
Latex Free Yes
Sterility Non-sterile
Waist Style Wide Elastic
For Use With (Application) Pharmaceutical, Lab, Healthcare, Controlled Environments
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.