Learn More
Braintree Scientific Soft Temp Reusale Veterinary Pad, 20 x 29, single pad
SDP
Catalog No. 501954200
Encompass
Soft-Temp Veterinary Heating Pads Soft-Temp reusable pads are the perfect solution for veterinary warming. Available in 4 sizes, the pads fit table-tops, surgery tables and in kennel cages. Soft-Temp pads work best with Adroits HTP-1500 pump, but adapters are available should you have a different pump. Pads may be used directly against the skin or covered with pillow cases or light-weight fabric for added protection against damage. They are heavy-duty and can be patched if needed. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4200 Supplier Braintree Scientific Supplier No. APAV029
Soft-Temp Veterinary Heating Pads Soft-Temp reusable pads are the perfect solution for veterinary warming. Available in 4 sizes, the pads fit table-tops, surgery tables and in kennel cages. Soft-Temp pads work best with Adroits HTP-1500 pump, but adapters are available should you have a different pump. Pads may be used directly against the skin or covered with pillow cases or light-weight fabric for added protection against damage. They are heavy-duty and can be patched if needed. **Sold for research purposes only and not for human use**