Learn More
Description
- Cost-effective solution
- <1% of the cost of stainless steel probes
- Significantly reduces cross contamination and associated risks
- Popular for use with low-analyte detection assays, such as PCR
- Commonly used for RNA and DNA extraction
- Simple 2-piece construction
- Compatible with Omni TH, THQ homogenizers and LH96 automation
Specifications
Specifications
| Product Type | Probe |
| Capacity (Metric) | 10 to 100 mg |
| Diameter (Metric) Probe | 5 mm |
| Dimensions (L x W x H) | 5 x 7 mm |
| Autoclavable | Autoclavable |
| Blade Material | Polypropylene |
| Material | Polycarbonate, Polyetherimide |
| Bottom Type | Flat |
| Closure Color | Clear |
| Color | Blue |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.