Learn More
US Biomax Inc Soft tissue sarcoma as control, including TNM, clinical stage and pathology grade, 80 cases/80 cores(core size 1.5mm), replacing SO809b
SDP
Catalog No. NC3433578 Shop All US Biomax Inc Products
Encompass
Soft tissue sarcoma microarray, containing 20 cases of liposarcoma, 18 fibrosarcoma, 2 UPS,20 leiomyosarcoma, plus 20 rhabdomyosarcoma, single core per case
Catalog No. NC3433578 Supplier US Biomax Inc Supplier No. SO809L32
Soft tissue sarcoma microarray, containing 20 cases of liposarcoma, 18 fibrosarcoma, 2 UPS,20 leiomyosarcoma, plus 20 rhabdomyosarcoma, single core per case