Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ SoftCIDE™ -ES Antimicrobial Handwash, bag-in-a-box carton refill

Catalog No. p-4525636 Shop All Epredia Products
Change view
Click to view available options
Volume (English):
27 oz.
32 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
12-544-302 32 oz.
12-544-301 27 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-544-302 Supplier Epredia™ Supplier No. 2803212001
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Effectively kill bacteria with Epredia™ SoftCIDE™-ES Antimicrobial Handwash, while still treating your skin gently.

  • Works against a broad spectrum of microorganisms
  • Formulated with emollients to help soften skin
  • Kills bacteria while it cleans
  • Offers high sudsing power and a thick, rich texture
  • pH balanced with the skin

Specifications

Product Type Anti-microbial Hand Wash
Description Pump bottle
Packaging Pump Bottle
Quantity 12/Cs.
Volume (English) 32 oz.
Volume (Metric) 946 mL
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.