Learn More
Research Products International Corp SoftCIDE Soap, 32 oz. Pump Bottle
SDP
Catalog No. 50212386
Encompass
SoftCIDE is specially designed for up to 40 washings per day without being harsh on your skin. SoftCIDE's unique formulation includes only high quality ingredients that are non-irritating. SoftCIDE contains 1% Chloroxylenol (PCMX) a proven anti-bacterial agent that is effective against bacteria, fungi and yeast.
WARNING: This product can expose you to chemicals which are known to the State of California to cause cancer and birth defects or other reproductive harm. For more information, go to www.p65Warnings.ca.gov.
Catalog No. 50-212-386 Supplier Research Products International Corp Supplier No. 151040
Add to Cart
Add to Cart
SoftCIDE is specially designed for up to 40 washings per day without being harsh on your skin. SoftCIDE's unique formulation includes only high quality ingredients that are non-irritating. SoftCIDE contains 1% Chloroxylenol (PCMX) a proven anti-bacterial agent that is effective against bacteria, fungi and yeast.
WARNING: This product can expose you to chemicals which are known to the State of California to cause cancer and birth defects or other reproductive harm. For more information, go to www.p65Warnings.ca.gov.