Learn More
Werner Co. SoftCoil Single Leg Lanyard
Catalog No. 17103819
Change view
Click to view available options
Material:
Polyester
Catalog No. Material
17-103-819 Polyester
1 options
Catalog No. 17-103-819 Supplier Werner Co. Supplier No. C351100
  • $78.00
    $72.65 / Each of 1
    Save $5.35
    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
SoftCoil single leg lanyard with energy absorbing inner core and snap hook.
The Werner SoftCoil lanyard specially designed inner core absorbs fall arrest energy along the entire length of the lanyard.
  • Integrated impact indicator gives user clear visual warning when the lanyard has been subjected to a fall and should be removed from service.
  • Snap hook is designed to prevent false engagement when connecting to the harness.
Specifications
Product Type SoftCoil Lanyard
Connector Types Snap Hook
Material Polyester
Leg Configuration Single
Webbing Material Internal Energy Absorbing Web
Length (English) 72 in.
Load Bearing Capacity (Metric) 1.633 kg Gate Strength
Length (Metric) 182.88 cm
Load Bearing Capacity (English) 3,600 lb. Gate Strength
Color Gold