Learn More
Hach Company High-Capacity, Deluxe Softener with Resin
Supplier:  Hach Company 3050001
Encompass
Large 1600-grain water softener. Capacity nearly double that of most softeners. For use with AD-16E Test Kit (Prod. No. 148410). 11.5" Height.
Catalog No. 50-199-7017