Learn More
Thermo Scientific™ SoftFit-L™ Empty Racks for use with SoftFit-L Reload Inserts and Reload System Towers
Shop All Thermo Scientific Products

Description
Thermo Scientific™ART SoftFit-L™ empty racks, when used in conjunction with SoftFit-L reload towers and inserts, offer an environmentally friendly packaging option with less plastic waste than traditional racks. Compatible with ART SoftFit-L reload inserts and reload towers.
Specifications
Specifications
| Volume (Metric) | 1000 μL |
| Model | SoftFit L Rack |
| Clean Claims | DNase, RNase, DNA, ATP, and Endotoxin Free |
| Format | Hinged Rack |
| Quantity | 8 Racks/Pk., 4 Pks./Cs. |
| For Use With (Equipment) | Rainin LTS LiteTouch |
| Product Line | ART SoftFit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.