Learn More
Thermo Scientific™ SoftFit-L™ Empty Racks for use with SoftFit-L Reload Inserts and Reload System Towers
GREENER_CHOICE
Catalog No. p-8459010 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
20 μL Pipet Tip, 200 μL Pipet Tip, 300 μL Pipet Tip
Rainin LTS LiteTouch
Quantity:
10 Racks/Pk., 5 Pks./Cs.
8 Racks/Pk., 4 Pks./Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Quantity
21-402-560 20 μL Pipet Tip, 200 μL Pipet Tip, 300 μL Pipet Tip 10 Racks/Pk., 5 Pks./Cs.
21-402-591 Rainin LTS LiteTouch 8 Racks/Pk., 4 Pks./Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-402-560 Supplier Thermo Scientific™ Supplier No. 3704ER
Add to Cart
Add to Cart
Thermo Scientific™ART SoftFit-L™ empty racks, when used in conjunction with SoftFit-L reload towers and inserts, offer an environmentally friendly packaging option with less plastic waste than traditional racks. Compatible with ART SoftFit-L reload inserts and reload towers.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Model SoftFit L Rack
Quantity 10 Racks/Pk., 5 Pks./Cs.
Product Type Pipet Tip Rack
For Use With (Equipment) 20 μL Pipet Tip, 200 μL Pipet Tip, 300 μL Pipet Tip
Product Line ART SoftFit