Learn More
Thermo Scientific™ SoftFit-L™ Pipette Tips, TipCycle Recycle Kit, Hinged Racks
GREENER_CHOICE
Catalog No. 02681563
Change view
Click to view available options
Volume (Metric):
20 μL
1000 μL
Sterility:
Non-sterile
Sterile
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility
02-681-563 20 μL Sterile
02-681-564 20 μL Non-sterile
02-681-571 1000 μL Sterile
02-681-572 1000 μL Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 02-681-563 Supplier Thermo Scientific™ Supplier No. 3721RCHR
Only null left
Add to Cart
Add to Cart
The pipette tip and rack mail-back recycling program provides a simple and effective solution for recycling single use plastics. The TipCycle Recycle Kit arrives with your case of pipette tips to collect used pipette tips and empty racks. Attach the pre-paid shipping label and return for recycling.
Gain the accuracy and precision you expect from your pipette samples every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
  • Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
  • Complete TipCycle Pipette Tip Recycling Kit Includes: 1 case of pipette tips, box liner, liner tie, FedEx return shipping label, waiver form, and instructions for return
  • Hinged rack provides easy one-hand access to tips
  • Color-coded inserts allow quick identification of tip volume
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 20 μL
Filtered Non-filtered
Sterility Sterile
Color Natural
Autoclavable Not Autoclavable
Model 20 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit
Length (English) 1.95 in.
Length (Metric) 4.86 cm
Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Format Hinged Rack
Tip Style Specialty
Product Type TipCycle, Pipette Tip and Rack Recycling Kit
Includes 1 Case of Pipette Tips, Recycle Box Liner, Liner Tie, Return Shipping Label, Waiver Form, and Instructions
Color Code Red
Product Line Softfit-L