Learn More
Thermo Scientific™ SoftFit-L™ Pipette Tips, TipCycle Recycle Kit, Hinged Racks
Shop All Thermo Scientific Products

Description
Gain the accuracy and precision you expect from your pipette samples every time with Thermo Scientific™ SoftFit-L™ pipette tips. For use with Rainin™ LTS pipettes.
- Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
- Complete TipCycle Pipette Tip Recycling Kit Includes: 1 case of pipette tips, box liner, liner tie, FedEx return shipping label, waiver form, and instructions for return
- Hinged rack provides easy one-hand access to tips
- Color-coded inserts allow quick identification of tip volume
Specifications
Specifications
| Volume (Metric) | 1000 μL |
| Filtered | Non-filtered |
| Sterility | Sterile |
| Color | Natural |
| Autoclavable | Not Autoclavable |
| Model | 1000 μL, Non-Filtered, Sterile, Hinged Rack, TipCycle Recycle Kit |
| Length (English) | 3.39 in. |
| Length (Metric) | 8.61 cm |
| Clean Claims | DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free |
| Format | Hinged Rack |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.