Learn More
Moldex™ Softies™ Ecostation™ Ear Plug Refill
SDP
Catalog No. 19384370 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Softies™ Ecostation™ Ear Plug Refill
Catalog No.
19-384-370 Softies™ Ecostation™ Ear Plug Refill
1 options
Catalog No. 19-384-370 Supplier Moldex™ Supplier No. 6702
Add to Cart
Add to Cart
Convenient refill container for Softies™ Ecostation™ dispenser (6701). Softies™ disposable ear plugs are made of extra-soft, extra-light foam allowing a gentle seal and comfortable fit.
  • Radiant orange color for easy compliance checks
  • Extra-soft, low-pressure foam for comfort and fit
  • Tapered shape for easy insertion and removal
  • Shaped to fit even very small ear canals
Specifications
ANSI S3.1974 Yes
CSA Z94.2 Class AL
Size Universal
For Use With (Application) Construction, Utilities, Manufacturing, Production, Mining, Oil and Gas, and Other Various Industries
High Visibility Yes
Packaging Type Refill
Pairs per Package 500 Pairs/Dispenser Refill
Plug Material Foam
Plug Color Orange
Noise Reduction Rating (NRR) 33 dB
Corded Uncorded
Plug Shape Bullet
Disposable Single-use
Metal Detectable No