Learn More
MSC Soft Scrub All-Purpose Cleaner, Disinfectant, Liquid, Unscented, 36 oz
Catalog No. 502870612
Encompass
This product is an all-purpose liquid cleaner designed for disinfecting hard surfaces. It comes in a 36 oz bottle and is unscented, suitable for various cleaning applications.
  • Disinfectant for hard surfaces
  • Unscented liquid formulation
  • EPA Safer Choice certified
  • Non-acidic for diverse applications
  • Packaged in a 36 oz bottle
Catalog No. 50-287-0612 Supplier MSC Supplier No. 09339292
This product is an all-purpose liquid cleaner designed for disinfecting hard surfaces. It comes in a 36 oz bottle and is unscented, suitable for various cleaning applications.
  • Disinfectant for hard surfaces
  • Unscented liquid formulation
  • EPA Safer Choice certified
  • Non-acidic for diverse applications
  • Packaged in a 36 oz bottle