Learn More
Medicus Health Red Specimen Transport Insulated Softside Tote, Shoulder Strap, Large
SDP
Catalog No. NC3252443
Encompass
Soft-Sided Specimen Transport Tote with embroidered biohazard symbol, shoulder strap and handle, large size is an easy and convenient way to transport specimens
• Specimen Cooler is perfect for transporting specimens to and from the lab while maintaining temperatures
• Heavy-duty, leak-proof 3/4" high density polyurethane foam insulated liner offers superior protection for your specimens or blood products
• Durable leak-resistant insulated liner is easy to clean and disinfect
• External Dimensions: 21"W x 10"D x 12"H; Light-weight and easy to transport
• Soft-sided cooler with high-density insulation, a removable shoulder strap, and side pocket
• Twice the insulation of other soft-sided transport coolers
• Made of durable soft canvas material that stands up to travel; features a high-quality leak-proof TPU liner
• Convenient transport handle and padded should strap with heavy-duty zipper to secure contents
Catalog No. NC3252443 Supplier Medicus Health Supplier No. 2936M3RED
Soft-Sided Specimen Transport Tote with embroidered biohazard symbol, shoulder strap and handle, large size is an easy and convenient way to transport specimens
• Specimen Cooler is perfect for transporting specimens to and from the lab while maintaining temperatures
• Heavy-duty, leak-proof 3/4" high density polyurethane foam insulated liner offers superior protection for your specimens or blood products
• Durable leak-resistant insulated liner is easy to clean and disinfect
• External Dimensions: 21"W x 10"D x 12"H; Light-weight and easy to transport
• Soft-sided cooler with high-density insulation, a removable shoulder strap, and side pocket
• Twice the insulation of other soft-sided transport coolers
• Made of durable soft canvas material that stands up to travel; features a high-quality leak-proof TPU liner
• Convenient transport handle and padded should strap with heavy-duty zipper to secure contents