Learn More
Thoren Caging SOFTTOUCH FORCEPS FOR MOUSE HO
SDP
Catalog No. NC0428970
Encompass
Thoren's SoftTouch Forceps are designed specifically for handling mice. Our engineers combined design and materials that would provide the correct force to pick-up a mouse eliminating any discomfort or pain to the mouse. No matter how much pressure you apply, only the force required to pick up the mouse is transfered. Quite amazing, and imagine, no more squeaking mice with SoftTouch Forceps. 10 per pack.
Catalog No. NC0428970 Supplier Thoren Caging Supplier No. ACFORECEPSU1
Thoren's SoftTouch Forceps are designed specifically for handling mice. Our engineers combined design and materials that would provide the correct force to pick-up a mouse eliminating any discomfort or pain to the mouse. No matter how much pressure you apply, only the force required to pick up the mouse is transfered. Quite amazing, and imagine, no more squeaking mice with SoftTouch Forceps. 10 per pack.