Learn More
Fritsch Milling And Sizing Inc Software Autosieve for Windows XP, Windows 7 ff.
SDP
Catalog No. 501533892
Encompass
The multilingual AUTOSIEVE programme for automatic evaluation, simple monitoring and documentation of your sieving results is delivered with each FRITSCH Sieve Shaker free of charge as a test version for 90 days. The software AUTOSIEVE is supplied on a practical USB stick and is therefore very quickly installed. All you have to do is to connect a laboratory analysis balance, and then weigh the sieves before and after sieving using differential weighing. AUTOSIEVE will do the rest. The software, which is suitable for Windows 7, XP and Vista, enables simple saving and retrieving of empty sieve weights, both of single sieves and complete sieve sets.
Catalog No. 50-153-3892 Supplier Fritsch Milling And Sizing Inc Supplier No. 03.2902.00
The multilingual AUTOSIEVE programme for automatic evaluation, simple monitoring and documentation of your sieving results is delivered with each FRITSCH Sieve Shaker free of charge as a test version for 90 days. The software AUTOSIEVE is supplied on a practical USB stick and is therefore very quickly installed. All you have to do is to connect a laboratory analysis balance, and then weigh the sieves before and after sieving using differential weighing. AUTOSIEVE will do the rest. The software, which is suitable for Windows 7, XP and Vista, enables simple saving and retrieving of empty sieve weights, both of single sieves and complete sieve sets.