Learn More
METTLER TOLEDO 3P INDUSTRIAL UV/VIS instrument license, Express Edition
Catalog No. NC1748137
Encompass_Preferred
Full version of LabX for installation on a single computer. Up to 3 instruments can be connected to this edition.
This item is not returnable. View return policy
Catalog No. NC1748137 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 30247984
Full version of LabX for installation on a single computer. Up to 3 instruments can be connected to this edition.