Learn More
Brady™ Markware™ Software and Laminating Kit
Catalog No. 19112936 Shop All Brady Corporation Products
Change view
Click to view available options
Software and Laminating Kit
Catalog No.
19-112-936 Software and Laminating Kit
1 options
Catalog No. 19-112-936 Supplier Brady™ Supplier No. 20711
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Covers all safety and facility identification needs in one powerful package
  • To design professional looking signs and labels using PC
  • Prints directly to labeling system or Windows inkjet/laser printers
  • Preformatted templates provided for quick and easy label creation
Specifications
Product Type Software and Laminating Kit
For Use With (Equipment) BMP33 Printer, BMP31 Printer, GlobalMark Printer, PowerMark Sign and Label Printer, MiniMark Printer, BMP71 Label Printer, HandiMark Printer
Languages Danish, Dutch, English, Finnish, French, German, Italian, Norwegian, Portuguese, Spanish, Swedish
Operating System Windows™ 2000 / XPSP2 / Vista
For Use With (Application) Energy Source Labeling, Facility, Safety and Maintenance Indentificaiton, Hazardous Material Labeling, NFPA Compliant, Production/ Bar Code Labeling Software, Rating Plate Identification, Right to Know Labeling, Visual Equipment Controls, Warehouse Bin and Shelf Labeling
Quantity 1 Ea.