Learn More
Aviva Systems Biology Corp SOHLH1 Antibody - middle region (ARP36303_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36303_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SOHLH1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SOHLH1. Alternate names: C9orf157, NOHLH, TEB2, bA100C15.3, bHLHe80ApplicationWB. Species reactivity: Human
Catalog No. 50-014-56110