Learn More
Aviva Systems Biology Corp Sohlh1 Antibody - middle region (ARP37844_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37844_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Sohlh1. It was validated on Western Blot. Sohlh1,spermatogenesis and oogenesis specific basic helix-loop-helix 1,Gm110; Nohlh; RP23-123F7.2; Sohlh1,spermatogenesis- and oogenesis-specific basic helix-loop-helix-containing protein 1,spermatogenesis- and oogenesis-specific basic helix-loop-helix-containing protein 1,OTTMUSP00000013510,helix-loop-helix protein NOHLHThe immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Gm110, Nohlh, RP23-123F7.2ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-284-689