Learn More
Aviva Systems Biology Corp SOHLH2 Antibody - middle region (ARP39313_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39313_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SOHLH2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SOHLH2. Alternate names: FLJ20449, TEB1, bA121N13.2, bHLHe81ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Rabbit
Catalog No. 50-014-56980