Learn More
Gilson Company Inc Soil Grinder (115V / 60Hz)
Catalog No. NC1326898
Encompass_Preferred
SA-45 Soil Grinder quickly prepares samples for Atterberg Limits or grain size testing and other standard lab tests. It is an efficient method for reducing agglomerations of caked soil to individual grains, and much less labor intensive than a manual mortar and pestle operation. The SA-45 grinders preserve true grain size for accurate and repeatable test results. The hopper has a 1pint (0.6L) capacity and features a manually operated gate to control feed rate to the grinding chamber.
Catalog No. NC1326898 Supplier Gilson Company Inc Supplier No. SA45
SA-45 Soil Grinder quickly prepares samples for Atterberg Limits or grain size testing and other standard lab tests. It is an efficient method for reducing agglomerations of caked soil to individual grains, and much less labor intensive than a manual mortar and pestle operation. The SA-45 grinders preserve true grain size for accurate and repeatable test results. The hopper has a 1pint (0.6L) capacity and features a manually operated gate to control feed rate to the grinding chamber.