Learn More
Andwin Scientific SOIL GRINDING KIT
SDP
Catalog No. 501125985
Encompass_Preferred
The Soil Grinding Kit is made for convenient lysing procedures with microorganisms living in soil, sediment, or roots. This kit comes with 50 preps, ready-to-use Precellys24, Precellys Evolution, or Minilys homogenizers.
Catalog No. 50-112-5985 Supplier Andwin Scientific Supplier No. 10144506
The Soil Grinding Kit is made for convenient lysing procedures with microorganisms living in soil, sediment, or roots. This kit comes with 50 preps, ready-to-use Precellys24, Precellys Evolution, or Minilys homogenizers.