Learn More
MSC NoTrax Soil Guard 340 Entrance Mat, Natural Rubber, 1 ea
Catalog No. 5000768140
Encompass
Soil Guard™ entrance mats are designed for effective soil and debris removal in high-traffic outdoor and indoor entryways. Made from a specially formulated rubber, these mats withstand harsh weather and heavy foot traffic, offering a low profile for easy door clearance and cart movement.
  • Heavy-duty, durable rubber compound
  • High-low design scrapes debris from shoe traffic
  • Low profile for door clearance and cart traffic
  • Easy to clean by hosing off
  • Wheelchair and cart friendly design
  • Beveled edges create a retention dam for moisture and debris
  • Ideal for high-traffic outdoor and indoor entryways
  • Constructed from natural rubber
  • Features raised studs surface detail
  • Passes US fire rating
Catalog No. 50-007-68140 Supplier MSC Supplier No. 05000534
Soil Guard™ entrance mats are designed for effective soil and debris removal in high-traffic outdoor and indoor entryways. Made from a specially formulated rubber, these mats withstand harsh weather and heavy foot traffic, offering a low profile for easy door clearance and cart movement.
  • Heavy-duty, durable rubber compound
  • High-low design scrapes debris from shoe traffic
  • Low profile for door clearance and cart traffic
  • Easy to clean by hosing off
  • Wheelchair and cart friendly design
  • Beveled edges create a retention dam for moisture and debris
  • Ideal for high-traffic outdoor and indoor entryways
  • Constructed from natural rubber
  • Features raised studs surface detail
  • Passes US fire rating