Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science Soil Macronutrient Testing Kit
GREENER_CHOICESCI_ED

Catalog No. S139198 Shop All Aldon Products
Change view
Click to view available options
Class Size:
50 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S139198 50 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139198 Supplier Innovating Science Supplier No. IS9761
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Plants need a variety of nutrients and minerals to grow and maintain overall health. Extract nutrients from soil of your choice and determine if nitrogen, phosphorus, and potassium are present in the soil sample.

  • Kit contains enough materials to perform 50 tests

Specifications

Grade 5+
Includes Instructional manual and all necessary chemicals
Class Size 50 Students
Product Type Educational Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.