Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Soil pH Meter
SCI_ED

Catalog No. S45153 Shop All Luster Leaf Products, Inc. Products
Change view
Click to view available options
For Use With (Application):
pH
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S45153 pH
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S45153 Supplier Luster Leaf Products, Inc. Supplier No. 1840 /6EA PER PK

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Meter designed exclusively for quick soil testing.

  • Measures soil pH from 3.5 to 9 so you can ensure proper plant nutrition
  • No more litmus paper, chemicals or messy solutions
  • No batteries required — meter works by sensing minute soil currents
  • Packaging includes detailed instruction with optimal pH values for more than 400 plants

Specifications

Includes Detailed instruction with optimal pH values for more than 400 plants
For Use With (Application) pH
Detection Range 3.5 to 9 pH
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.