Learn More
BURKLE INC Soil sampler GeoSampler, 81 cm, flute length 11.81 in. (300 mm)
Catalog No. 5000025667
Encompass_Preferred
Soil sampler for light soils, meadows and lawns. The foot bar makes work much easier.
Catalog No. 50-000-25667 Supplier BURKLE INC Supplier No. 53505006
Soil sampler for light soils, meadows and lawns. The foot bar makes work much easier.