Learn More
BURKLE INC Soil sampler Puerckhauer, 46.26 in. (117,5 cm), flute 39.37 in. (100 cm)
Catalog No. 5000025621
Encompass_Preferred
Heavy-duty Puerckhauer soil sampler designed for tough, rocky soils. Featuring a conically tapered tip for easy extraction after driving into the ground, this sampler ensures efficient and reliable soil sampling in challenging conditions. The durable construction is perfect for taking N-min samples at depths ranging from 0 to 90 cm (0 to 35.4 inches).
Available in various lengths with flute sizes up to 100 cm (39.37 inches), this soil sampler delivers accurate and consistent samples for agricultural, environmental, and research applications. Ideal for professionals seeking a robust and efficient tool for soil analysis.
Catalog No. 50-000-25621 Supplier BURKLE INC Supplier No. 53505018
Heavy-duty Puerckhauer soil sampler designed for tough, rocky soils. Featuring a conically tapered tip for easy extraction after driving into the ground, this sampler ensures efficient and reliable soil sampling in challenging conditions. The durable construction is perfect for taking N-min samples at depths ranging from 0 to 90 cm (0 to 35.4 inches).
Available in various lengths with flute sizes up to 100 cm (39.37 inches), this soil sampler delivers accurate and consistent samples for agricultural, environmental, and research applications. Ideal for professionals seeking a robust and efficient tool for soil analysis.