Learn More
Cole-Parmer Soil Sampling Sieve Set, Stainless Steel, 2" Height
Supplier:  Cole-Parmer 9902699
Encompass_Preferred
Wildco 78-700 Soil Sampling Sieve Set, Stainless Steel, 2" Height, Great for separating and grading rock and soil samplesSix of the most commonly used U.S.standard sizesStackable, INCLUDESSix sieves [U.S. Standard No. 5 (4000 micron), U.S. Standard No. 10 (2000 micron), U.S. Standard No. 35 (500 micron), U.S. Standard No. 60 (250 micron), U.S. Standard No. 120 (125 micron), U.S. Standard No. 230 (63 micron)], lid, and catch pan
Catalog No. NC1498243