Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Soil Test Kit
SCI_ED

Catalog No. s17702 Shop All Luster Leaf Products, Inc. Products
Change view
Click to view available options
Product Type:
Soil Test Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S17702 Soil Test Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S17702 Supplier Luster Leaf Products, Inc. Supplier No. 1601

Accurate test results within the hour.

  • Measure soil's pH, nitrogen, phosphorus and potassium — up to 10 times each
  • Color comparator system
  • Color-coded reagent capsules perform 40 total tests

Specifications

For Use With (Application) Measure the soil ph, nitrogen, phosphorous and potassium
Includes Easy-to-use capsule system, 4 individual color comparators, ph and npk levels chart and complete instructions
Time Range 1
Dimensions (L x W x H) W: 9.13 in.; H: 8.5 in.; D: 1.31 in.
Product Type Soil Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.